Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152903.4 | NP_690867.3 | 1 | 1852 | ||
GenBank mRNA | AK056633.1 | - | 1 | 1797 | ||
AK095195.1 | - | 1 | 497 | |||
AK096608.1 | - | 2 | 1792 | |||
AK299627.1 | - | 2 | 1600 | |||
AL833839.1 | - | 1 | 1504 | |||
BC000560.2 | - | 1 | 1774 | 3163484 | ||
BC051349.1 | - | 1 | 1613 | 6483483 |