Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LDB2 SIRNA Assays ›
Gene Symbol:
LDB2
Gene Name:
LIM domain binding 2
Gene Aliases:
CLIM1, LDB-2, LDB1
Chromosome Location:
Chr.4: 16501534 - 16898809 on Build GRCh38
UniGene ID:
Hs.714330
Species:
Human
siRNA ID
HSS189843
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LDB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130834.2
NP_001124306.1
9
1422
NM_001290.4
NP_001281.1
8
1279
NM_001304434.1
NP_001291363.1
8
1273
NM_001304435.1
NP_001291364.1
8
1537
XM_005248197.3
XP_005248254.1
8
1246
XM_005248201.2
XP_005248258.1
9
1389
XM_005248202.2
XP_005248259.1
9
1386
XM_006713975.3
XP_006714038.1
10
1308
XM_006713976.3
XP_006714039.1
10
1306
XM_006713977.3
XP_006714040.1
10
1302
XM_006713978.3
XP_006714041.1
11
1452
XM_006713979.3
XP_006714042.1
11
1710
XM_006713980.3
XP_006714043.1
11
1450
XM_006713981.3
XP_006714044.1
11
1446
XM_006713982.3
XP_006714045.1
10
1567
XM_006713983.3
XP_006714046.1
10
1561
XM_011513905.2
XP_011512207.1
9
1200
XM_017008812.1
XP_016864301.1
10
1300
XM_017008813.1
XP_016864302.1
8
1240
XM_017008814.1
XP_016864303.1
11
1444
XM_017008815.1
XP_016864304.1
10
1565
XM_017008816.1
XP_016864305.1
10
1559
XM_017008817.1
XP_016864306.1
9
1647
XM_017008818.1
XP_016864307.1
8
846
XM_017008819.1
XP_016864308.1
8
1504
XM_017008820.1
XP_016864309.1
8
1501
XM_017008821.1
XP_016864310.1
8
1498
XM_017008822.1
XP_016864311.1
9
1189
GenBank mRNA
AF047337.1
-
8
1064
AF052389.1
-
9
1131
AF064492.1
-
8
1064
AF064493.1
-
9
1206
AF068651.1
-
8
1062
AK095238.1
-
2
1452
AK097013.1
-
1
1339
AK226029.1
-
8
1362
AK294588.1
-
7
1092
AK300119.1
-
8
870
AK312881.1
-
9
1117
AL832772.1
-
1
5072
BC034019.1
-
8
1072
4693654
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
hair follicle development
epithelial structure maintenance
regulation of cell migration
somatic stem cell population maintenance
regulation of kinase activity
positive regulation of cellular component biogenesis
positive regulation of transcription from RNA polymerase II promoter
transcription cofactor activity
enzyme binding
LIM domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qvdz4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0