Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033273.2 | NP_150376.1 | 4, 5 | 537 | ||
XM_011515604.2 | XP_011513906.1 | 5, 6 | 723 | |||
XM_011515608.2 | XP_011513910.1 | 5, 6 | 627 | |||
XM_011515609.2 | XP_011513911.1 | 3, 4 | 518 | |||
XM_011515610.2 | XP_011513912.1 | 6, 7 | 804 | |||
XM_011515611.2 | XP_011513913.1 | 4, 5 | 671 | |||
XM_017012777.1 | XP_016868266.1 | 3, 4 | 372 | |||
XM_017012778.1 | XP_016868267.1 | 4, 5 | 575 | |||
GenBank mRNA | AF277624.1 | - | 4, 5 | 537 |