Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130726.2 | NP_001124198.1 | 7 | 794 | ||
NM_173463.4 | NP_775734.2 | 8 | 895 | |||
XM_005248210.3 | XP_005248267.1 | 7 | 823 | |||
XM_011513906.2 | XP_011512208.1 | 9 | 905 | |||
XM_011513907.2 | XP_011512209.1 | 8 | 778 | |||
XM_011513908.1 | XP_011512210.1 | 7 | 700 | |||
XM_017008827.1 | XP_016864316.1 | 9 | 905 | |||
XM_017008828.1 | XP_016864317.1 | 8 | 829 | |||
GenBank mRNA | AK125353.1 | - | 7 | 703 | ||
AK296454.1 | - | 4 | 452 | |||
AK302905.1 | - | 8 | 839 | |||
AL834257.2 | - | 7 | 768 | |||
DR423861.1 | - | 7 | 709 |