Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD1E SIRNA Assays ›
Gene Symbol:
CD1E
Gene Name:
CD1e molecule
Gene Aliases:
CD1A, R2
Chromosome Location:
Chr.1: 158353157 - 158357554 on Build GRCh38
UniGene ID:
Hs.249217
Species:
Human
siRNA ID
HSS189929
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CD1E
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042583.2
NP_001036048.1
5
1256
NM_001042584.2
NP_001036049.1
5
1152
NM_001042585.2
NP_001036050.1
5
1091
NM_001042586.2
NP_001036051.1
3
689
NM_001042587.2
NP_001036052.1
3
585
NM_001185107.1
NP_001172036.1
4
986
NM_001185108.1
NP_001172037.1
4
821
NM_001185110.1
NP_001172039.1
3
524
NM_001185112.1
NP_001172041.1
3
689
NM_001185113.1
NP_001172042.1
3
524
NM_001185114.1
NP_001172043.1
4
959
NM_001185115.1
NP_001172044.1
4
986
NM_030893.3
NP_112155.2
5
1256
XM_011510133.2
XP_011508435.1
5
1630
XM_011510134.2
XP_011508436.1
5
1691
GenBank mRNA
AJ289111.1
-
5
963
AJ289112.1
-
5
963
AJ289113.1
-
5
859
AJ289114.1
-
5
798
AJ289115.1
-
4
693
AJ289116.1
-
4
693
AJ289117.1
-
4
528
AJ289118.1
-
3
396
AJ289119.1
-
3
396
AJ289120.1
-
3
292
AJ289121.1
-
3
231
AJ289122.1
-
3
231
AK303088.1
-
4
1360
AK303108.1
-
5
1058
AK311643.1
-
4
875
BC131693.1
-
5
967
40120328
X79517.1
-
2
99
X79519.1
-
1
55
Back To Top
More Information
Panther Classification:
Molecular Function -
major histocompatibility complex protein
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
immune response
antigen processing and presentation, exogenous lipid antigen via MHC class Ib
beta-2-microglobulin binding
endogenous lipid antigen binding
exogenous lipid antigen binding
lipopeptide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0