Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKDCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138370.2 | NP_612379.2 | 3 | 1001 | ||
GenBank mRNA | AK311155.1 | - | 1 | 564 | ||
BC094697.1 | - | 3 | 409 | 30529646 | ||
BC110513.1 | - | 3 | 438 | 40031755 | ||
BC110514.1 | - | 3 | 438 | 40031756 | ||
CF131963.1 | - | 3 | 497 |