Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WDR20 SIRNA Assays ›
Gene Symbol:
WDR20
Gene Name:
WD repeat domain 20
Gene Aliases:
DMR
Chromosome Location:
Chr.14: 102139518 - 102224686 on Build GRCh38
UniGene ID:
Hs.36859
Species:
Human
siRNA ID
HSS189998
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WDR20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242414.1
NP_001229343.1
3
1458
NM_001242416.1
NP_001229345.1
2
1445
NM_001242417.1
NP_001229346.1
4
1721
NM_001242418.1
NP_001229347.1
3
1664
NM_144574.3
NP_653175.2
3
1628
NM_181291.2
NP_851808.1
3
1628
NM_181308.2
NP_851825.1
2
1445
NR_038354.1
4
1421
XM_006720308.1
XP_006720371.1
2
1437
XM_006720309.1
XP_006720372.1
3
1411
XM_006720310.1
XP_006720373.1
3
1372
XM_006720311.1
XP_006720374.1
2
1201
XM_006720313.2
XP_006720376.1
4
1380
XM_006720318.2
XP_006720381.1
3
1319
XM_011537335.2
XP_011535637.1
4
1740
XM_011537336.2
XP_011535638.1
4
1740
XM_011537337.2
XP_011535639.1
4
1740
XM_011537338.1
XP_011535640.1
4
1687
XM_011537339.2
XP_011535641.1
4
1740
XM_011537340.2
XP_011535642.1
4
1740
XM_011537341.2
XP_011535643.1
3
1646
XM_011537343.1
XP_011535645.1
5
1715
XM_011537344.2
XP_011535646.1
4
1633
XM_011537345.1
XP_011535647.1
3
1411
XM_011537346.1
XP_011535648.1
3
1437
XM_011537347.1
XP_011535649.1
4
1435
XM_011537353.2
XP_011535655.1
4
1491
XM_011537358.2
XP_011535660.1
3
1396
XM_017021766.1
XP_016877255.1
4
1687
XM_017021767.1
XP_016877256.1
3
1646
XM_017021768.1
XP_016877257.1
3
1661
XM_017021770.1
XP_016877259.1
3
1661
XM_017021771.1
XP_016877260.1
4
1696
XM_017021772.1
XP_016877261.1
4
1449
XM_017021773.1
XP_016877262.1
5
1542
XM_017021774.1
XP_016877263.1
4
1491
XM_017021775.1
XP_016877264.1
4
1497
XM_017021776.1
XP_016877265.1
3
1411
XM_017021777.1
XP_016877266.1
3
1379
XM_017021778.1
XP_016877267.1
4
1491
XM_017021779.1
XP_016877268.1
2
1201
XM_017021780.1
XP_016877269.1
4
1612
XM_017021781.1
XP_016877270.1
3
1319
XM_017021782.1
XP_016877271.1
3
1132
XM_017021783.1
XP_016877272.1
4
1428
XM_017021785.1
XP_016877274.1
4
1687
GenBank mRNA
AB209148.1
-
1
5717
AF327354.1
-
3
1553
AK056330.1
-
3
1370
AK090580.1
-
4
1380
AK090978.1
-
3
1399
AK225852.1
-
2
1404
AK297727.1
-
4
1659
AL133558.1
-
1
865
AL832242.1
-
3
1397
AL832500.1
-
3
1603
BC028387.2
-
3
1587
4823662
BX248274.1
-
2
1347
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f2nf4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0