Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WDR20 SIRNA Assays ›
Gene Symbol:
WDR20
Gene Name:
WD repeat domain 20
Gene Aliases:
DMR
Chromosome Location:
Chr.14: 102139518 - 102224686 on Build GRCh38
UniGene ID:
Hs.36859
Species:
Human
siRNA ID
HSS189999
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WDR20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242414.1
NP_001229343.1
3
777
NM_001242416.1
NP_001229345.1
2
764
NM_001242417.1
NP_001229346.1
4
1040
NM_001242418.1
NP_001229347.1
3
983
NM_144574.3
NP_653175.2
3
947
NM_181291.2
NP_851808.1
3
947
NM_181308.2
NP_851825.1
2
764
NR_038354.1
4
740
XM_006720308.1
XP_006720371.1
2
756
XM_006720309.1
XP_006720372.1
3
730
XM_006720310.1
XP_006720373.1
3
691
XM_006720311.1
XP_006720374.1
2
520
XM_006720313.2
XP_006720376.1
4
699
XM_006720318.2
XP_006720381.1
3
638
XM_011537335.2
XP_011535637.1
4
1059
XM_011537336.2
XP_011535638.1
4
1059
XM_011537337.2
XP_011535639.1
4
1059
XM_011537338.1
XP_011535640.1
4
1006
XM_011537339.2
XP_011535641.1
4
1059
XM_011537340.2
XP_011535642.1
4
1059
XM_011537341.2
XP_011535643.1
3
965
XM_011537343.1
XP_011535645.1
5
1034
XM_011537344.2
XP_011535646.1
4
952
XM_011537345.1
XP_011535647.1
3
730
XM_011537346.1
XP_011535648.1
3
756
XM_011537347.1
XP_011535649.1
4
754
XM_011537353.2
XP_011535655.1
4
810
XM_011537358.2
XP_011535660.1
3
715
XM_017021766.1
XP_016877255.1
4
1006
XM_017021767.1
XP_016877256.1
3
965
XM_017021768.1
XP_016877257.1
3
980
XM_017021770.1
XP_016877259.1
3
980
XM_017021771.1
XP_016877260.1
4
1015
XM_017021772.1
XP_016877261.1
4
768
XM_017021773.1
XP_016877262.1
5
861
XM_017021774.1
XP_016877263.1
4
810
XM_017021775.1
XP_016877264.1
4
816
XM_017021776.1
XP_016877265.1
3
730
XM_017021777.1
XP_016877266.1
3
698
XM_017021778.1
XP_016877267.1
4
810
XM_017021779.1
XP_016877268.1
2
520
XM_017021780.1
XP_016877269.1
4
931
XM_017021781.1
XP_016877270.1
3
638
XM_017021782.1
XP_016877271.1
3
451
XM_017021783.1
XP_016877272.1
4
747
XM_017021785.1
XP_016877274.1
4
1006
GenBank mRNA
AB209148.1
-
1
5036
AF327354.1
-
3
872
AK056330.1
-
3
689
AK090580.1
-
4
699
AK090978.1
-
3
718
AK225852.1
-
2
723
AK297727.1
-
4
978
AL133558.1
-
1
184
AL832242.1
-
3
716
AL832500.1
-
3
922
BC028387.2
-
3
906
4823662
BX248274.1
-
2
666
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0