Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052859.3 | NP_443091.1 | 5 | 528 | ||
XM_005265537.4 | XP_005265594.1 | 5 | 541 | |||
XM_006713384.3 | XP_006713447.1 | 5 | 539 | |||
XM_011534214.2 | XP_011532516.1 | 5 | 543 | |||
XM_011534215.2 | XP_011532517.1 | 5 | 543 | |||
XM_011534216.2 | XP_011532518.1 | 4 | 443 | |||
XM_017007460.1 | XP_016862949.1 | 5 | 540 | |||
XM_017007461.1 | XP_016862950.1 | 5 | 529 | |||
GenBank mRNA | AJ318099.1 | - | 5 | 512 | ||
BC043595.1 | - | 5 | 521 | 5769824 |