Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144575.2 | NP_653176.2 | 11 | 1282 | ||
XM_011533159.2 | XP_011531461.1 | 11 | 1279 | |||
XM_011533160.2 | XP_011531462.1 | 12 | 1470 | |||
XM_011533161.2 | XP_011531463.1 | 11 | 1279 | |||
XM_011533163.2 | XP_011531465.1 | 11 | 1279 | |||
XM_017005264.1 | XP_016860753.1 | 12 | 1470 | |||
XM_017005265.1 | XP_016860754.1 | 11 | 1279 | |||
XM_017005266.1 | XP_016860755.1 | 11 | 1279 | |||
GenBank mRNA | AK074418.1 | - | 12 | 1349 | ||
AK290298.1 | - | 11 | 1283 | |||
AK308037.1 | - | 11 | 1259 | |||
BC117345.1 | - | 11 | 1243 | 40125896 | ||
BC117347.1 | - | 11 | 1243 | 40125898 | ||
BX640793.1 | - | 11 | 1282 | |||
BX647678.1 | - | 4 | 377 |