Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012631.1 | NP_001012649.1 | 7 | 450 | ||
NM_001012632.1 | NP_001012650.1 | 6 | 249 | |||
NM_001012633.1 | NP_001012651.1 | 6 | 383 | |||
NM_001012634.1 | NP_001012652.1 | 5 | 190 | |||
NM_001012635.1 | NP_001012653.1 | 5 | 185 | |||
NM_001012636.1 | NP_001012654.1 | 4 | 173 | |||
NM_001012718.1 | NP_001012736.1 | 6 | 386 | |||
NM_001308078.1 | NP_001295007.1 | 5 | 384 | |||
NM_004221.4 | NP_004212.4 | 6 | 383 | |||
GenBank mRNA | AK296143.1 | - | 5 | 195 | ||
AK310053.1 | - | 3 | 926 | |||
AY495331.1 | - | 6 | 373 | |||
AY495332.1 | - | 6 | 373 | |||
AY495333.1 | - | 5 | 331 | |||
BC009401.2 | - | 5 | 521 | 4300407 | ||
BC018782.1 | - | 6 | 230 | 4873608 | ||
BC105602.1 | - | 6 | 251 | 6496101 | ||
BF568456.1 | - | 5 | 522 | |||
BG748458.1 | - | 6 | 329 | |||
BM759359.1 | - | 3 | 124 | |||
BM822755.1 | - | 7 | 358 | |||
JN546102.1 | - | 4 | 112 | |||
M59807.1 | - | 5 | 369 |