Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004224.3 | NP_004215.2 | 2 | 390 | ||
GenBank mRNA | AY114101.1 | - | 2 | 102 | ||
BC103696.1 | - | 2 | 300 | 40006713 | ||
BC105683.1 | - | 2 | 300 | 40006718 | ||
BC105684.1 | - | 2 | 300 | 40006715 | ||
BP245442.1 | - | 2 | 390 | |||
EU432118.1 | - | 2 | 252 | |||
U52219.1 | - | 2 | 321 |
Set Membership: |
![]() |