Gene Symbol: TIMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001563.2 | NP_001001563.1 | 8 | 1123 | ||
XM_011527491.2 | XP_011525793.1 | 8 | 666 | |||
XM_017027472.1 | XP_016882961.1 | 7 | 630 | |||
GenBank mRNA | AK098197.1 | - | 2 | 785 | ||
AY444561.1 | - | 7 | 598 | |||
AY551341.1 | - | 8 | 681 | |||
BC009072.1 | - | 8 | 655 | |||
BC010736.1 | - | 8 | 676 | |||
BC050082.1 | - | 8 | 699 | |||
BC121146.1 | - | 7 | 992 | 40121256 | ||
BC121147.1 | - | 8 | 1051 | 40121258 | ||
BQ925812.1 | - | 8 | 710 |