Gene Symbol: TIMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001563.2 | NP_001001563.1 | 6, 7 | 942 | ||
XM_011527491.2 | XP_011525793.1 | 6, 7 | 485 | |||
XM_017027472.1 | XP_016882961.1 | 5, 6 | 449 | |||
GenBank mRNA | AY444561.1 | - | 5, 6 | 417 | ||
AY551341.1 | - | 6, 7 | 500 | |||
BC009072.1 | - | 6, 7 | 474 | |||
BC010736.1 | - | 6, 7 | 495 | |||
BC050082.1 | - | 6, 7 | 518 | |||
BC121146.1 | - | 5, 6 | 811 | 40121256 | ||
BC121147.1 | - | 6, 7 | 870 | 40121258 | ||
BQ925812.1 | - | 6, 7 | 529 |