Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADGRG1 SIRNA Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
BFPP, BPPR, GPR56, TM7LN4, TM7XN1
Chromosome Location:
Chr.16: 57619535 - 57665039 on Build GRCh38
UniGene ID:
Hs.513633
Species:
Human
siRNA ID
HSS190149
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADGRG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145770.2
NP_001139242.1
14
1977
NM_001145771.2
NP_001139243.1
14
2433
NM_001145772.2
NP_001139244.1
14
2441
NM_001145773.2
NP_001139245.1
13
2312
NM_001145774.2
NP_001139246.1
14
2214
NM_001290142.1
NP_001277071.1
13
1535
NM_001290143.1
NP_001277072.1
13
1884
NM_001290144.1
NP_001277073.1
13
2053
NM_005682.6
NP_005673.3
14
1995
NM_201524.3
NP_958932.1
14
2419
NM_201525.3
NP_958933.1
13
2297
XM_005256237.2
XP_005256294.1
13
2031
XM_005256238.2
XP_005256295.1
14
2370
XM_005256239.2
XP_005256296.1
14
2132
XM_005256240.4
XP_005256297.1
13
1853
XM_005256241.4
XP_005256298.1
13
1857
XM_005256242.4
XP_005256299.1
13
1914
XM_005256244.3
XP_005256301.1
13
2006
XM_005256245.2
XP_005256302.1
13
2057
XM_005256246.2
XP_005256303.1
14
2117
XM_005256247.2
XP_005256304.1
13
2016
XM_005256248.2
XP_005256305.1
13
2081
XM_005256249.3
XP_005256306.1
13
1872
XM_005256251.4
XP_005256308.1
13
1898
XM_005256252.2
XP_005256309.1
13
1842
XM_005256254.2
XP_005256311.1
13
2042
XM_005256255.2
XP_005256312.1
14
2337
XM_006721338.2
XP_006721401.1
14
2153
XM_006721339.3
XP_006721402.1
14
1960
XM_006721340.3
XP_006721403.1
14
2218
XM_006721341.2
XP_006721404.1
14
2175
XM_006721342.2
XP_006721405.1
14
2138
XM_006721343.3
XP_006721406.1
14
2203
XM_006721344.2
XP_006721407.1
14
2160
XM_006721345.3
XP_006721408.1
14
1929
XM_006721346.2
XP_006721409.1
13
1784
XM_006721347.2
XP_006721410.1
14
2352
XM_011523461.2
XP_011521763.1
14
2121
XM_011523462.2
XP_011521764.1
13
1936
XM_011523463.2
XP_011521765.1
14
1946
XM_011523464.2
XP_011521766.1
14
2106
XM_011523465.2
XP_011521767.1
14
2044
XM_011523466.2
XP_011521768.1
13
2091
XM_011523467.2
XP_011521769.1
14
1838
XM_011523468.2
XP_011521770.1
13
1840
XM_017023892.1
XP_016879381.1
13
2024
GenBank mRNA
AF106858.1
-
13
1865
AJ011001.1
-
14
2001
AK075270.1
-
14
1929
AK075555.1
-
14
1952
AK131550.1
-
13
1490
AK297236.1
-
13
1926
AK299110.1
-
13
1841
AK311156.1
-
3
271
AK312405.1
-
13
2133
AY358400.1
-
14
1916
BC008770.2
-
13
1861
3139174
BC013207.1
-
14
1991
BT007311.1
-
12
1703
CR936747.1
-
13
1826
EU432119.1
-
12
1703
KU178472.1
-
12
1703
KU178473.1
-
12
1685
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cell adhesion
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
Rho protein signal transduction
cell-cell signaling
brain development
negative regulation of cell proliferation
vascular endothelial growth factor production
cell migration
cerebral cortex regionalization
cerebral cortex radial glia guided migration
layer formation in cerebral cortex
positive regulation of Rho protein signal transduction
positive regulation of cell adhesion
protein kinase C signaling
positive regulation of neural precursor cell proliferation
negative regulation of neuron migration
G-protein coupled receptor activity
collagen binding
heparin binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0