Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099337.1 | NP_001092807.1 | 4 | 555 | ||
NM_144576.3 | NP_653177.3 | 4 | 799 | |||
GenBank mRNA | AK057003.1 | - | 4 | 715 | ||
AY358861.1 | - | 4 | 487 | |||
BC002435.2 | - | 3 | 376 | |||
BC026922.1 | - | 4 | 555 | |||
BC047444.1 | - | 4 | 527 | |||
BC073923.1 | - | 4 | 683 | 5188138 |