Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ITM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004867.4 | NP_004858.1 | 2 | 493 | ||
GenBank mRNA | AB209310.1 | - | 1 | 3788 | ||
AF038953.1 | - | 2 | 295 | |||
AK223327.1 | - | 2 | 272 | |||
AK313154.1 | - | 2 | 451 | |||
AY359051.1 | - | 2 | 234 | |||
BC040437.1 | - | 2 | 248 | 5748519 | ||
BG506587.1 | - | 2 | 492 | |||
CR456875.1 | - | 2 | 156 | |||
DA365726.1 | - | 2 | 537 |