Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004867.4 | NP_004858.1 | 2, 3 | 586 | ||
GenBank mRNA | AF038953.1 | - | 2, 3 | 388 | ||
AK223327.1 | - | 2, 3 | 365 | |||
AK313154.1 | - | 2, 3 | 544 | |||
AY359051.1 | - | 2, 3 | 327 | |||
BC040437.1 | - | 2, 3 | 341 | 5748519 | ||
BG506587.1 | - | 2, 3 | 585 | |||
CR456875.1 | - | 2, 3 | 249 |