Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039477.2 | NP_001034566.1 | 2 | 212 | ||
NM_001105556.2 | NP_001099026.1 | 2 | 212 | |||
NM_001286113.1 | NP_001273042.1 | 2 | 212 | |||
NM_001286115.1 | NP_001273044.1 | 2 | 212 | |||
NM_004848.3 | NP_004839.2 | 2 | 212 | |||
XM_005246041.3 | XP_005246098.1 | 2 | 224 | |||
XM_006711050.1 | XP_006711113.1 | 2 | 206 | |||
GenBank mRNA | AK303090.1 | - | 2 | 174 | ||
AK303141.1 | - | 2 | 208 | |||
AL554503.3 | - | 2 | 176 | |||
BC031655.1 | - | 2 | 180 | |||
BC132692.1 | - | 2 | 211 | |||
BC133049.1 | - | 2 | 211 | |||
BG716895.1 | - | 2 | 205 | |||
BM146655.1 | - | 2 | 212 | |||
CR749321.1 | - | 2 | 213 |