Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOCS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014011.4 | NP_054730.1 | 2 | 1281 | ||
NM_144949.2 | NP_659198.1 | 2 | 1369 | |||
GenBank mRNA | AB014571.1 | - | 2 | 1289 | ||
AF073958.1 | - | 2 | 1369 | |||
AK290194.1 | - | 2 | 1295 | |||
AK296796.1 | - | 2 | 1239 | |||
AL136896.1 | - | 2 | 1281 | |||
BC032862.1 | - | 2 | 1382 | 5264754 |