Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE4DIP SIRNA Assays ›
Gene Symbol:
PDE4DIP
LOC100996724
Gene Name:
phosphodiesterase 4D interacting protein
phosphodiesterase 4D interacting protein-like
Gene Aliases:
CMYA2, MMGL
-
Chromosome Location:
Chr.1: 148808167 - 149051277 on Build GRCh38
Chromosome Location:
Chr.1: 120471449 - 120701470 on Build GRCh38
UniGene ID:
Hs.584841
Hs.731111
Species:
Human
siRNA ID
HSS190507
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE4DIP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002812.2
NP_001002812.2
4, 5
824
NM_001198832.2
NP_001185761.2
7, 8
1106
NM_001198834.3
NP_001185763.3
4, 5
824
NM_014644.5
NP_055459.5
4, 5
824
XM_006711646.1
XP_006711709.1
7, 8
1113
XM_006711650.3
XP_006711713.1
5, 6
598
XM_006711651.1
XP_006711714.1
4, 5
716
XM_006711652.1
XP_006711715.1
4, 5
716
XM_011510172.1
XP_011508474.1
7, 8
1271
XM_011510173.1
XP_011508475.1
7, 8
1271
XM_011510175.1
XP_011508477.1
7, 8
1271
XM_011510176.1
XP_011508478.1
7, 8
1271
XM_011510179.1
XP_011508481.1
4, 5
716
XM_017002878.1
XP_016858367.1
7, 8
1271
XM_017002880.1
XP_016858369.1
7, 8
1271
XM_017002887.1
XP_016858376.1
8, 9
1548
XM_017002888.1
XP_016858377.1
8, 9
1521
XM_017002889.1
XP_016858378.1
8, 9
1193
XM_017002891.1
XP_016858380.1
6, 7
1352
XM_017002892.1
XP_016858381.1
6, 7
983
XM_017002893.1
XP_016858382.1
7, 8
1471
XM_017002894.1
XP_016858383.1
7, 8
1116
XM_017002895.1
XP_016858384.1
4, 5
434
XM_017002898.1
XP_016858387.1
7, 8
1113
XM_017002899.1
XP_016858388.1
7, 8
1113
XM_017002902.1
XP_016858391.1
7, 8
1113
XM_017002903.1
XP_016858392.1
7, 8
1113
GenBank mRNA
AB007923.2
-
7, 8
985
AL831815.2
-
4, 5
667
AL832024.2
-
4, 5
632
AL833273.1
-
4, 5
712
BX647660.1
-
4, 5
752
Gene Symbol: LOC100996724
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017003064.1
XP_016858553.1
7, 8
1267
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein complex assembly
protein binding
enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline