Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIMS2 SIRNA Assays ›
Gene Symbol:
RIMS2
Gene Name:
regulating synaptic membrane exocytosis 2
Gene Aliases:
OBOE, RAB3IP3, RIM2
Chromosome Location:
Chr.8: 103500026 - 104254430 on Build GRCh38
UniGene ID:
Hs.655271
Species:
Human
siRNA ID
HSS190539
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RIMS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100117.2
NP_001093587.1
4
1745
NM_001282881.1
NP_001269810.1
2
1350
NM_014677.4
NP_055492.3
2
1350
XM_005251106.3
XP_005251163.1
4
1745
XM_005251107.3
XP_005251164.1
4
1745
XM_011517395.2
XP_011515697.1
8
1798
XM_011517398.2
XP_011515700.1
8
1798
XM_017014006.1
XP_016869495.1
8
1798
XM_017014007.1
XP_016869496.1
8
1798
XM_017014008.1
XP_016869497.1
8
1798
XM_017014009.1
XP_016869498.1
8
1798
XM_017014010.1
XP_016869499.1
8
1798
XM_017014011.1
XP_016869500.1
8
1798
XM_017014012.1
XP_016869501.1
8
1798
XM_017014013.1
XP_016869502.1
7
1756
XM_017014014.1
XP_016869503.1
4
1745
XM_017014015.1
XP_016869504.1
8
1798
XM_017014016.1
XP_016869505.1
8
1798
XM_017014017.1
XP_016869506.1
8
1798
XM_017014018.1
XP_016869507.1
4
1745
XM_017014019.1
XP_016869508.1
6
1877
XM_017014020.1
XP_016869509.1
5
2312
XM_017014021.1
XP_016869510.1
8
1798
XM_017014022.1
XP_016869511.1
2
1269
XM_017014023.1
XP_016869512.1
8
1798
XM_017014024.1
XP_016869513.1
8
1798
XM_017014025.1
XP_016869514.1
8
1798
XM_017014026.1
XP_016869515.1
6
2018
XM_017014027.1
XP_016869516.1
8
1798
XM_017014028.1
XP_016869517.1
8
1798
XM_017014029.1
XP_016869518.1
5
2479
XM_017014030.1
XP_016869519.1
4
1745
XM_017014031.1
XP_016869520.1
8
1798
XM_017014032.1
XP_016869521.1
8
1798
XM_017014033.1
XP_016869522.1
7
2598
XM_017014034.1
XP_016869523.1
5
2479
XM_017014035.1
XP_016869524.1
5
2478
XM_017014036.1
XP_016869525.1
2
1269
XM_017014037.1
XP_016869526.1
5
2478
XM_017014038.1
XP_016869527.1
4
1745
XM_017014039.1
XP_016869528.1
7
2599
XM_017014040.1
XP_016869529.1
2
1269
XM_017014041.1
XP_016869530.1
2
1269
XM_017014042.1
XP_016869531.1
4
1745
XM_017014043.1
XP_016869532.1
2
1269
XM_017014044.1
XP_016869533.1
2
1269
XM_017014045.1
XP_016869534.1
2
1269
XM_017014046.1
XP_016869535.1
2
1269
GenBank mRNA
AB018294.1
-
2
1278
AK126939.1
-
4
1745
AK307790.1
-
2
1316
AK308831.1
-
2
1150
BC018057.1
-
2
1151
BC043144.2
-
2
1266
5288275
CR749374.1
-
2
1182
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
positive regulation of gene expression
calcium ion regulated exocytosis
regulation of exocytosis
cAMP-mediated signaling
insulin secretion
regulation of membrane potential
calcium ion-regulated exocytosis of neurotransmitter
spontaneous neurotransmitter secretion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
Rab GTPase binding
ion channel binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0