Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST18 SIRNA Assays ›
Gene Symbol:
ST18
Gene Name:
ST18, C2H2C-type zinc finger
Gene Aliases:
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location:
Chr.8: 52110832 - 52409879 on Build GRCh38
UniGene ID:
Hs.655499
Species:
Human
siRNA ID
HSS190546
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014682.2
NP_055497.1
10
1325
XM_006716487.1
XP_006716550.1
10
1092
XM_011517629.1
XP_011515931.1
11
1248
XM_011517631.1
XP_011515933.1
12
1289
XM_011517632.1
XP_011515934.1
10
1103
XM_011517633.1
XP_011515935.1
10
1096
XM_011517634.1
XP_011515936.1
10
1292
XM_011517635.1
XP_011515937.1
10
1296
XM_011517636.2
XP_011515938.1
9
1373
XM_011517637.1
XP_011515939.1
8
830
XM_011517638.2
XP_011515940.1
7
871
XM_011517641.1
XP_011515943.1
12
1336
XM_011517642.1
XP_011515944.1
11
1248
XM_017014047.1
XP_016869536.1
10
1452
XM_017014048.1
XP_016869537.1
10
1456
XM_017014049.1
XP_016869538.1
11
1502
XM_017014050.1
XP_016869539.1
10
1289
XM_017014051.1
XP_016869540.1
9
1296
XM_017014052.1
XP_016869541.1
11
1506
XM_017014053.1
XP_016869542.1
11
1493
XM_017014054.1
XP_016869543.1
9
1542
XM_017014055.1
XP_016869544.1
9
1133
XM_017014056.1
XP_016869545.1
12
1543
XM_017014057.1
XP_016869546.1
10
1346
XM_017014058.1
XP_016869547.1
10
1107
XM_017014059.1
XP_016869548.1
10
1293
XM_017014060.1
XP_016869549.1
11
1330
XM_017014061.1
XP_016869550.1
9
1465
XM_017014062.1
XP_016869551.1
8
1217
XM_017014063.1
XP_016869552.1
9
1763
XM_017014064.1
XP_016869553.1
9
1377
XM_017014065.1
XP_016869554.1
11
1497
XM_017014066.1
XP_016869555.1
9
1136
XM_017014067.1
XP_016869556.1
9
1546
XM_017014068.1
XP_016869557.1
10
1337
XM_017014069.1
XP_016869558.1
7
1034
XM_017014070.1
XP_016869559.1
6
955
XM_017014071.1
XP_016869560.1
7
1203
XM_017014072.1
XP_016869561.1
10
1454
XM_017014073.1
XP_016869562.1
10
1092
XM_017014074.1
XP_016869563.1
9
1375
XM_017014075.1
XP_016869564.1
11
1539
XM_017014076.1
XP_016869565.1
11
1543
XM_017014077.1
XP_016869566.1
11
1180
XM_017014078.1
XP_016869567.1
10
1460
XM_017014079.1
XP_016869568.1
10
1851
XM_017014080.1
XP_016869569.1
8
1092
XM_017014081.1
XP_016869570.1
10
1459
XM_017014082.1
XP_016869571.1
10
1294
XM_017014083.1
XP_016869572.1
9
1134
XM_017014084.1
XP_016869573.1
7
872
GenBank mRNA
AB011107.1
-
10
1189
BC031075.1
-
10
1222
BC117147.1
-
5
582
40125698
BC117149.1
-
5
582
40125700
BC143872.1
-
5
577
BC143873.1
-
6
670
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0