Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM94 SIRNA Assays ›
Gene Symbol:
TMEM94
Gene Name:
transmembrane protein 94
Gene Aliases:
KIAA0195
Chromosome Location:
Chr.17: 75441159 - 75500452 on Build GRCh38
UniGene ID:
Hs.514474
Species:
Human
siRNA ID
HSS190614
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM94
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321148.1
NP_001308077.1
18
2745
NM_001321149.1
NP_001308078.1
19
2929
NM_014738.5
NP_055553.3
19
2917
XM_005257864.2
XP_005257921.1
18
2752
XM_011525514.1
XP_011523816.1
19
2840
XM_011525515.1
XP_011523817.1
20
2966
XM_011525516.1
XP_011523818.1
19
2756
XM_011525517.1
XP_011523819.1
19
2735
XM_011525519.1
XP_011523821.1
13
1896
XM_011525520.2
XP_011523822.1
16
2396
XM_011525521.1
XP_011523823.1
6
1056
XM_017025433.1
XP_016880922.1
18
2782
XM_017025434.1
XP_016880923.1
18
2779
XM_017025435.1
XP_016880924.1
18
2770
XM_017025436.1
XP_016880925.1
18
2752
XM_017025437.1
XP_016880926.1
18
2740
XM_017025438.1
XP_016880927.1
18
2734
XM_017025439.1
XP_016880928.1
18
2722
XM_017025440.1
XP_016880929.1
19
2798
XM_017025441.1
XP_016880930.1
18
2782
XM_017025442.1
XP_016880931.1
18
2782
XM_017025443.1
XP_016880932.1
18
2692
XM_017025444.1
XP_016880933.1
18
2779
XM_017025445.1
XP_016880934.1
18
2737
XM_017025446.1
XP_016880935.1
18
2704
XM_017025447.1
XP_016880936.1
18
2782
XM_017025448.1
XP_016880937.1
18
2692
XM_017025449.1
XP_016880938.1
19
2750
XM_017025450.1
XP_016880939.1
17
2560
GenBank mRNA
AK128508.1
-
11
1556
AK294526.1
-
18
2708
AK304819.1
-
11
1692
AK316060.1
-
18
2745
BC045540.1
-
19
2686
5274473
CR936809.1
-
16
3291
D83779.1
-
19
2726
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0