Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014765.2 | NP_055580.1 | 4, 5 | 613 | ||
GenBank mRNA | AK289810.1 | - | 4, 5 | 613 | ||
BC000882.1 | - | 4, 5 | 505 | 3462090 | ||
BC066335.1 | - | 4, 5 | 501 | 4818590 | ||
BC071994.1 | - | 4, 5 | 484 | 6082526 | ||
BC100286.1 | - | 4, 5 | 495 | 6157036 | ||
BC107851.1 | - | 4, 5 | 491 | 5579754 | ||
CR456718.1 | - | 4, 5 | 387 | |||
D13641.1 | - | 4, 5 | 488 |