Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP170 SIRNA Assays ›
Gene Symbol:
CEP170
Gene Name:
centrosomal protein 170
Gene Aliases:
FAM68A, KAB, KIAA0470
Chromosome Location:
Chr.1: 243124428 - 243255936 on Build GRCh38
UniGene ID:
Hs.533635
Species:
Human
siRNA ID
HSS190671
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP170
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042404.1
NP_001035863.1
12
2442
NM_001042405.1
NP_001035864.1
12
2442
NM_014812.2
NP_055627.2
13
2736
XM_006711843.3
XP_006711906.1
12
3445
XM_011544334.2
XP_011542636.1
12
3452
XM_011544335.2
XP_011542637.1
12
2865
XM_011544336.2
XP_011542638.1
12
3771
XM_011544337.2
XP_011542639.1
12
3452
XM_011544338.2
XP_011542640.1
12
3452
XM_011544339.2
XP_011542641.1
12
3452
XM_011544340.2
XP_011542642.1
12
3402
XM_011544341.2
XP_011542643.1
13
3216
XM_011544342.2
XP_011542644.1
12
3402
XM_011544343.2
XP_011542645.1
11
2925
XM_011544344.2
XP_011542646.1
12
2972
XM_017002932.1
XP_016858421.1
12
3365
XM_017002933.1
XP_016858422.1
13
3036
XM_017002934.1
XP_016858423.1
13
2679
XM_017002935.1
XP_016858424.1
13
3216
XM_017002936.1
XP_016858425.1
13
2679
XM_017002937.1
XP_016858426.1
13
3216
XM_017002938.1
XP_016858427.1
13
3216
XM_017002939.1
XP_016858428.1
13
3036
XM_017002940.1
XP_016858429.1
13
2679
XM_017002941.1
XP_016858430.1
13
3036
XM_017002942.1
XP_016858431.1
13
3216
XM_017002943.1
XP_016858432.1
13
3036
XM_017002944.1
XP_016858433.1
12
2965
XM_017002945.1
XP_016858434.1
12
2972
XM_017002946.1
XP_016858435.1
12
2739
XM_017002947.1
XP_016858436.1
12
2972
XM_017002948.1
XP_016858437.1
12
2965
XM_017002949.1
XP_016858438.1
12
2972
XM_017002950.1
XP_016858439.1
12
2739
XM_017002951.1
XP_016858440.1
11
2445
GenBank mRNA
AB007939.1
-
11
2087
AB022657.1
-
12
2381
AB022658.1
-
11
2087
AB022659.1
-
11
2087
AK001664.1
-
1
24
AK296812.1
-
12
2160
AK308309.1
-
2
533
BC140794.1
-
11
2078
9021662
BC146790.1
-
11
2087
9007254
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0