Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIM66 SIRNA Assays ›
Gene Symbol:
TRIM66
Gene Name:
tripartite motif containing 66
Gene Aliases:
C11orf29, TIF1D, TIF1DELTA
Chromosome Location:
Chr.11: 8612037 - 8683217 on Build GRCh38
UniGene ID:
Hs.130836
Species:
Human
siRNA ID
HSS190677
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIM66
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014818.1
NP_055633.1
8, 9
977
XM_006718397.2
XP_006718460.1
13, 14
1822
XM_006718398.3
XP_006718461.1
9, 10
2434
XM_011520504.1
XP_011518806.1
14, 15
1865
XM_011520507.2
XP_011518809.1
13, 14
1786
XM_011520508.1
XP_011518810.1
13, 14
1822
XM_011520509.1
XP_011518811.1
13, 14
1822
XM_011520510.1
XP_011518812.1
13, 14
1822
XM_011520511.1
XP_011518813.1
13, 14
1711
XM_011520512.1
XP_011518814.1
9, 10
1086
XM_011520513.1
XP_011518815.1
9, 10
1084
XM_011520514.2
XP_011518816.1
9, 10
1040
XM_011520515.1
XP_011518817.1
10, 11
1253
XM_011520516.1
XP_011518818.1
10, 11
1253
XM_011520517.2
XP_011518819.1
10, 11
2575
XM_011520518.1
XP_011518820.1
10, 11
1223
XM_011520519.1
XP_011518821.1
9, 10
1111
XM_011520522.2
XP_011518824.1
8, 9
2289
XM_011520523.2
XP_011518825.1
9, 10
3171
XM_011520524.1
XP_011518826.1
13, 14
1822
XM_011520525.1
XP_011518827.1
13, 14
1822
XM_017018629.1
XP_016874118.1
13, 14
2215
XM_017018630.1
XP_016874119.1
8, 9
1177
XM_017018631.1
XP_016874120.1
8, 9
1329
GenBank mRNA
AB002296.2
-
8, 9
977
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Process(es)
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0