Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133350.1 | NP_579928.1 | 4 | 430 | ||
GenBank mRNA | AF468005.1 | - | 4 | 430 | ||
AK177097.1 | - | 4 | 345 | |||
BC048833.1 | - | 3 | 992 | |||
BC057918.1 | - | 4 | 430 | 5134461 | ||
U51204.1 | - | 2 | 112 |