Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133350.1 | NP_579928.1 | 4, 5 | 607 | ||
GenBank mRNA | AF468005.1 | - | 4, 5 | 607 | ||
BC048833.1 | - | 3, 4 | 1169 | |||
BC057918.1 | - | 4, 5 | 607 | 5134461 | ||
U51204.1 | - | 2, 3 | 289 |