Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028990.4 | NP_083266.1 | 2 | 504 | ||
GenBank mRNA | AK034589.1 | - | 2 | 473 | ||
AK051532.1 | - | 2 | 505 | |||
AK054353.1 | - | 2 | 461 | |||
AK152957.1 | - | 2 | 468 | |||
AK170302.1 | - | 2 | 481 | |||
BC007160.1 | - | 2 | 448 | 3488466 | ||
BC024911.1 | - | 2 | 423 | 3594306 |