Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242349.1 | NP_001229278.1 | 14 | 1651 | ||
NM_178642.5 | NP_848757.4 | 15 | 1660 | |||
GenBank mRNA | AK029329.1 | - | 1 | 27 | ||
AK032793.1 | - | 10 | 672 | |||
AK052589.1 | - | 15 | 1612 | |||
BC004697.1 | - | 7 | 526 | |||
BC062959.1 | - | 14 | 1537 | 30547439 |