Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005863.2 | NP_001005863.1 | 9 | 3413 | ||
NM_001005864.3 | NP_001005864.1 | 7 | 1019 | |||
NM_001005865.3 | NP_001005865.2 | 5 | 1032 | |||
NM_001286413.1 | NP_001273342.1 | 9 | 3295 | |||
GenBank mRNA | AF173380.1 | - | 5 | 871 | ||
AF493235.1 | - | 5 | 936 | |||
AK030510.1 | - | 5 | 996 | |||
AK031693.1 | - | 5 | 966 | |||
AK035576.1 | - | 3 | 400 | |||
AK129324.1 | - | 4 | 2464 | |||
AY246699.1 | - | 5 | 694 | |||
AY626781.1 | - | 5 | 1029 | |||
AY626782.1 | - | 7 | 1017 | |||
AY626783.1 | - | 8 | 3235 | |||
BC041777.1 | - | 8 | 2078 | 4240274 | ||
BC042206.1 | - | 8 | 2078 | 4240274 | ||
BC043321.1 | - | 7 | 983 | |||
BC089009.1 | - | 9 | 3295 | 30543240 |