Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005863.2 | NP_001005863.1 | 8 | 3141 | ||
NM_001005864.3 | NP_001005864.1 | 6 | 747 | |||
NM_001005865.3 | NP_001005865.2 | 4 | 760 | |||
NM_001286413.1 | NP_001273342.1 | 8 | 3023 | |||
GenBank mRNA | AF173380.1 | - | 4 | 599 | ||
AF493235.1 | - | 4 | 664 | |||
AK030510.1 | - | 4 | 724 | |||
AK031693.1 | - | 4 | 694 | |||
AK035576.1 | - | 2 | 128 | |||
AK129324.1 | - | 4 | 746 | |||
AY246699.1 | - | 4 | 422 | |||
AY626781.1 | - | 4 | 757 | |||
AY626782.1 | - | 6 | 745 | |||
AY626783.1 | - | 7 | 2963 | |||
BC041777.1 | - | 7 | 1806 | 4240274 | ||
BC042206.1 | - | 7 | 1806 | 4240274 | ||
BC043321.1 | - | 6 | 711 | |||
BC089009.1 | - | 8 | 3023 | 30543240 |