Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018743.4 | NP_061213.2 | 2 | 1003 | ||
GenBank mRNA | AF406611.1 | - | 1 | 514 | ||
AK045235.1 | - | 2 | 897 | |||
AK049675.1 | - | 2 | 897 | |||
AK077561.1 | - | 2 | 887 | |||
AK083589.1 | - | 2 | 897 | |||
AK161270.1 | - | 2 | 866 | |||
AK167876.1 | - | 2 | 912 | |||
AK169257.1 | - | 2 | 897 | |||
AY489184.1 | - | 1 | 72 | |||
BC031767.1 | - | 1 | 357 | 4457846 |