Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271347.1 | NP_001258276.1 | 4, 5 | 899 | ||
NM_001271348.1 | NP_001258277.1 | 4, 5 | 860 | |||
NM_001271349.1 | NP_001258278.1 | 3, 4 | 797 | |||
NM_134015.3 | NP_598776.1 | 5, 6 | 962 | |||
GenBank mRNA | AB093260.1 | - | 3, 4 | 604 | ||
AK041532.1 | - | 4, 5 | 692 | |||
AK149139.1 | - | 4, 5 | 682 | |||
AK152181.1 | - | 4, 5 | 622 | |||
AK160086.1 | - | 3, 4 | 598 | |||
AK166226.1 | - | 4, 5 | 557 | |||
AK168667.1 | - | 3, 4 | 614 | |||
AK218380.1 | - | 2, 3 | 287 | |||
AY038079.1 | - | 5, 6 | 626 | |||
BC034261.1 | - | 5, 6 | 642 | 4237375 |