Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007596.2 | NP_001007597.1 | 4 | 600 | ||
NM_001286448.1 | NP_001273377.1 | 4 | 1145 | |||
NM_153457.7 | NP_703187.2 | 6 | 2681 | |||
GenBank mRNA | AB074899.1 | - | 6 | 2178 | ||
AK002997.1 | - | 4 | 598 | |||
AK013731.1 | - | 4 | 535 | |||
AK014358.1 | - | 4 | 553 | |||
AK132364.1 | - | 6 | 2681 | |||
BC027091.1 | - | 4 | 480 | |||
BC030455.1 | - | 6 | 2233 | 5401661 | ||
BC053926.1 | - | 6 | 2309 | 6312477 | ||
BC058579.1 | - | 6 | 2343 | 6490092 | ||
CT010392.1 | - | 6 | 2178 |