Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007596.2 | NP_001007597.1 | 6, 7 | 721 | ||
NM_001286448.1 | NP_001273377.1 | 6, 7 | 1266 | |||
NM_153457.7 | NP_703187.2 | 8, 9 | 2802 | |||
GenBank mRNA | AB074899.1 | - | 8, 9 | 2299 | ||
AK002997.1 | - | 6, 7 | 719 | |||
AK013731.1 | - | 6, 7 | 656 | |||
AK014358.1 | - | 6, 7 | 674 | |||
AK132364.1 | - | 8, 9 | 2802 | |||
BC027091.1 | - | 6, 7 | 601 | |||
BC030455.1 | - | 8, 9 | 2354 | 5401661 | ||
BC053926.1 | - | 8, 9 | 2430 | 6312477 | ||
BC058579.1 | - | 8, 9 | 2464 | 6490092 | ||
CT010392.1 | - | 8, 9 | 2299 |