Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113514.1 | NP_001106986.1 | 12 | 1254 | ||
NM_133721.2 | NP_598482.2 | 26 | 3075 | |||
GenBank mRNA | AJ344342.1 | - | 26 | 3078 | ||
AK083987.1 | - | 11 | 1094 | |||
AK139690.1 | - | 12 | 1242 | |||
AK153585.1 | - | 24 | 2537 | |||
BC049939.1 | - | 9 | 871 | |||
BC150824.1 | - | 26 | 2981 | 9087735 |