Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipas39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142580.1 | NP_001136052.1 | 16 | 1276 | ||
NM_001142581.1 | NP_001136053.1 | 15 | 1219 | |||
NM_134044.3 | NP_598805.2 | 15 | 1233 | |||
GenBank mRNA | AK046322.1 | - | 16 | 1247 | ||
AK049643.1 | - | 16 | 1223 | |||
AK079055.1 | - | 16 | 1223 | |||
AK082261.1 | - | 16 | 1204 | |||
BC023716.1 | - | 15 | 1244 | 5346489 |