Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289598.1 | NP_001276527.1 | 8 | 1032 | ||
NM_001289599.1 | NP_001276528.1 | 8 | 999 | |||
NM_145367.4 | NP_663342.3 | 8 | 1056 | |||
GenBank mRNA | AK136046.1 | - | 8 | 944 | ||
AK142049.1 | - | 8 | 1042 | |||
AK150781.1 | - | 8 | 1040 | |||
AK169680.1 | - | 8 | 1023 | |||
AK224911.1 | - | 7 | 709 | |||
AK224913.1 | - | 8 | 1043 | |||
AK224922.1 | - | 8 | 1001 | |||
AK224923.1 | - | 8 | 1023 | |||
AK224924.1 | - | 8 | 1031 | |||
AK224926.1 | - | 8 | 1001 | |||
AK224927.1 | - | 8 | 1052 | |||
AK224930.1 | - | 8 | 1004 | |||
AK224933.1 | - | 8 | 993 | |||
AK224934.1 | - | 8 | 1047 | |||
AK224935.1 | - | 1 | 1198 | |||
AK224936.1 | - | 8 | 1046 | |||
AK224937.1 | - | 8 | 1045 | |||
AK224938.1 | - | 8 | 1040 | |||
AK224939.1 | - | 8 | 1033 | |||
AK224943.1 | - | 8 | 1004 | |||
AK224945.1 | - | 8 | 1040 | |||
AK224946.1 | - | 8 | 1043 | |||
AK224947.1 | - | 8 | 1030 | |||
AK224948.1 | - | 8 | 1004 | |||
AK224949.1 | - | 8 | 993 | |||
AK224950.1 | - | 8 | 993 | |||
AK224951.1 | - | 8 | 1051 | |||
AK224955.1 | - | 3 | 304 | |||
AK224956.1 | - | 8 | 963 | |||
AK224957.1 | - | 8 | 1009 | |||
AK224958.1 | - | 8 | 1009 | |||
AK224960.1 | - | 8 | 1051 | |||
AK224962.1 | - | 8 | 1011 | |||
AK224965.1 | - | 8 | 1028 | |||
AK224966.1 | - | 8 | 1028 | |||
AK224967.1 | - | 8 | 1019 | |||
AK224968.1 | - | 6 | 586 | |||
AK224969.1 | - | 6 | 586 | |||
AK224970.1 | - | 8 | 993 | |||
AK224971.1 | - | 8 | 1053 | |||
AK224972.1 | - | 8 | 1043 | |||
AK224973.1 | - | 8 | 993 | |||
AK224974.1 | - | 8 | 996 | |||
AK224975.1 | - | 3 | 231 | |||
AK224976.1 | - | 8 | 1026 | |||
AK224977.1 | - | 8 | 1040 | |||
AK224978.1 | - | 8 | 1040 | |||
AK224979.1 | - | 8 | 1027 | |||
AK224981.1 | - | 8 | 997 | |||
AK224982.1 | - | 8 | 1052 | |||
AK224983.1 | - | 8 | 1041 | |||
AK224984.1 | - | 8 | 1006 | |||
AK224985.1 | - | 8 | 1010 | |||
AK224986.1 | - | 8 | 1054 | |||
AK224988.1 | - | 8 | 1030 | |||
AK224989.1 | - | 8 | 1047 | |||
AK224990.1 | - | 8 | 1042 | |||
AK224991.1 | - | 8 | 1037 | |||
AK224992.1 | - | 8 | 1037 | |||
AK224993.1 | - | 8 | 1037 | |||
AK224994.1 | - | 8 | 1045 | |||
AK224995.1 | - | 8 | 1008 | |||
AK224996.1 | - | 8 | 979 | |||
AK224997.1 | - | 8 | 1004 | |||
AK224998.1 | - | 8 | 1002 | |||
AK224999.1 | - | 8 | 1024 | |||
AK225004.1 | - | 8 | 1045 | |||
AK225005.1 | - | 8 | 1023 | |||
AK225007.1 | - | 8 | 1045 | |||
AK225010.1 | - | 8 | 1045 | |||
AK225011.1 | - | 8 | 1045 | |||
AK225012.1 | - | 8 | 814 | |||
AK225013.1 | - | 8 | 814 | |||
AK225014.1 | - | 8 | 1023 | |||
AK225015.1 | - | 8 | 814 | |||
AK225016.1 | - | 8 | 953 | |||
AK225017.1 | - | 8 | 1001 | |||
AK225019.1 | - | 8 | 1033 | |||
AK225020.1 | - | 8 | 984 | |||
AK225021.1 | - | 8 | 984 | |||
AK225022.1 | - | 8 | 832 | |||
AK225023.1 | - | 8 | 1058 | |||
AY243534.1 | - | 8 | 1019 | |||
AY548112.1 | - | 8 | 980 | |||
BC016252.1 | - | 8 | 1023 | 4911455 | ||
BC024505.1 | - | 8 | 968 | 4983496 | ||
BC036155.1 | - | 7 | 642 | |||
BC046789.1 | - | 8 | 982 | 5716897 |