Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289598.1 | NP_001276527.1 | 9, 10 | 1180 | ||
NM_001289599.1 | NP_001276528.1 | 9, 10 | 1147 | |||
NM_145367.4 | NP_663342.3 | 9, 10 | 1204 | |||
GenBank mRNA | AK142049.1 | - | 9, 10 | 1190 | ||
AK150781.1 | - | 9, 10 | 1188 | |||
AK169680.1 | - | 9, 10 | 1171 | |||
AK224911.1 | - | 8, 9 | 857 | |||
AK224913.1 | - | 9, 10 | 1191 | |||
AK224922.1 | - | 9, 10 | 1149 | |||
AK224923.1 | - | 9, 10 | 1171 | |||
AK224924.1 | - | 9, 10 | 1179 | |||
AK224926.1 | - | 9, 10 | 1149 | |||
AK224927.1 | - | 9, 10 | 1200 | |||
AK224930.1 | - | 9, 10 | 1152 | |||
AK224933.1 | - | 9, 10 | 1141 | |||
AK224934.1 | - | 9, 10 | 1195 | |||
AK224935.1 | - | 2, 3 | 1346 | |||
AK224936.1 | - | 9, 10 | 1194 | |||
AK224937.1 | - | 9, 10 | 1193 | |||
AK224938.1 | - | 9, 10 | 1188 | |||
AK224939.1 | - | 9, 10 | 1181 | |||
AK224943.1 | - | 9, 10 | 1152 | |||
AK224945.1 | - | 9, 10 | 1188 | |||
AK224946.1 | - | 9, 10 | 1191 | |||
AK224947.1 | - | 9, 10 | 1178 | |||
AK224948.1 | - | 9, 10 | 1152 | |||
AK224949.1 | - | 9, 10 | 1141 | |||
AK224950.1 | - | 9, 10 | 1141 | |||
AK224951.1 | - | 9, 10 | 1199 | |||
AK224955.1 | - | 4, 5 | 452 | |||
AK224956.1 | - | 9, 10 | 1111 | |||
AK224957.1 | - | 9, 10 | 1157 | |||
AK224958.1 | - | 9, 10 | 1157 | |||
AK224960.1 | - | 9, 10 | 1199 | |||
AK224962.1 | - | 9, 10 | 1159 | |||
AK224965.1 | - | 9, 10 | 1176 | |||
AK224966.1 | - | 9, 10 | 1176 | |||
AK224967.1 | - | 9, 10 | 1167 | |||
AK224968.1 | - | 7, 8 | 734 | |||
AK224969.1 | - | 7, 8 | 734 | |||
AK224970.1 | - | 9, 10 | 1141 | |||
AK224971.1 | - | 9, 10 | 1201 | |||
AK224972.1 | - | 9, 10 | 1191 | |||
AK224973.1 | - | 9, 10 | 1141 | |||
AK224974.1 | - | 9, 10 | 1144 | |||
AK224975.1 | - | 4, 5 | 379 | |||
AK224976.1 | - | 9, 10 | 1174 | |||
AK224977.1 | - | 9, 10 | 1188 | |||
AK224978.1 | - | 9, 10 | 1188 | |||
AK224979.1 | - | 9, 10 | 1175 | |||
AK224981.1 | - | 9, 10 | 1145 | |||
AK224982.1 | - | 9, 10 | 1200 | |||
AK224983.1 | - | 9, 10 | 1189 | |||
AK224984.1 | - | 9, 10 | 1154 | |||
AK224985.1 | - | 9, 10 | 1158 | |||
AK224986.1 | - | 9, 10 | 1202 | |||
AK224988.1 | - | 9, 10 | 1178 | |||
AK224989.1 | - | 9, 10 | 1195 | |||
AK224990.1 | - | 9, 10 | 1190 | |||
AK224991.1 | - | 9, 10 | 1185 | |||
AK224992.1 | - | 9, 10 | 1185 | |||
AK224993.1 | - | 9, 10 | 1185 | |||
AK224994.1 | - | 9, 10 | 1193 | |||
AK224995.1 | - | 9, 10 | 1156 | |||
AK224996.1 | - | 9, 10 | 1127 | |||
AK224997.1 | - | 9, 10 | 1152 | |||
AK224998.1 | - | 9, 10 | 1150 | |||
AK224999.1 | - | 9, 10 | 1172 | |||
AK225004.1 | - | 9, 10 | 1193 | |||
AK225005.1 | - | 9, 10 | 1171 | |||
AK225007.1 | - | 9, 10 | 1193 | |||
AK225010.1 | - | 9, 10 | 1193 | |||
AK225011.1 | - | 9, 10 | 1193 | |||
AK225012.1 | - | 9, 10 | 962 | |||
AK225013.1 | - | 9, 10 | 962 | |||
AK225015.1 | - | 9, 10 | 962 | |||
AK225016.1 | - | 9, 10 | 1101 | |||
AK225017.1 | - | 9, 10 | 1149 | |||
AK225019.1 | - | 9, 10 | 1181 | |||
AK225020.1 | - | 9, 10 | 1132 | |||
AK225021.1 | - | 9, 10 | 1132 | |||
AK225022.1 | - | 9, 10 | 980 | |||
AK225023.1 | - | 9, 10 | 1206 | |||
AY243534.1 | - | 9, 10 | 1167 | |||
AY548112.1 | - | 9, 10 | 1128 | |||
BC016252.1 | - | 9, 10 | 1171 | 4911455 | ||
BC024505.1 | - | 9, 10 | 1116 | 4983496 | ||
BC036155.1 | - | 8, 9 | 790 | |||
BC046789.1 | - | 9, 10 | 1130 | 5716897 |