Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134089.1 | NP_598850.1 | 30 | 4326 | ||
GenBank mRNA | AF271735.1 | - | 14 | 1820 | ||
AF441233.1 | - | 30 | 4326 | |||
AK122211.1 | - | 29 | 4296 | |||
BC006859.1 | - | 15 | 1896 | |||
BC037480.1 | - | 10 | 1148 | |||
BC049942.1 | - | 5 | 256 | |||
BC060689.1 | - | 22 | 3121 | 6417366 | ||
BC062888.1 | - | 29 | 4234 | 5721250 |