Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134091.2 | NP_598852.2 | 12 | 1472 | ||
GenBank mRNA | AK142314.1 | - | 12 | 1440 | ||
AK170806.1 | - | 12 | 1439 | |||
AK183939.1 | - | 3 | 291 | |||
AY382616.1 | - | 12 | 1426 | |||
BC018197.1 | - | 12 | 1432 | 4190407 | ||
BC024122.1 | - | 12 | 1432 | 5097970 | ||
BC025561.1 | - | 12 | 1410 | 5322980 |