Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Topors | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134097.3 | NP_598858.2 | 3 | 2264 | ||
GenBank mRNA | AB104865.1 | - | 3 | 2249 | ||
AK134075.1 | - | 3 | 2260 | |||
AK143025.1 | - | 3 | 2245 | |||
AK153743.1 | - | 3 | 2251 | |||
BC019421.1 | - | 3 | 2222 | |||
BC037141.1 | - | 3 | 2242 | |||
BC040797.1 | - | 3 | 2252 | 3492178 |