Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134136.3 | NP_598897.2 | 4 | 564 | ||
GenBank mRNA | AK031096.1 | - | 4 | 514 | ||
AK089046.1 | - | 4 | 536 | |||
AY267463.1 | - | 4 | 497 | |||
BC056348.1 | - | 4 | 516 | 5721407 |