Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025965.3 | NP_080241.3 | 4 | 614 | ||
GenBank mRNA | AF326229.2 | - | 4 | 594 | ||
AF395811.1 | - | 4 | 594 | |||
AK010884.1 | - | 4 | 560 | |||
AK031538.1 | - | 4 | 594 | |||
AK160285.1 | - | 4 | 577 | |||
AK167793.1 | - | 4 | 560 | |||
AK192546.1 | - | 1 | 113 | |||
AY993942.1 | - | 594 |