- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Usp9y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148943.2 | NP_683745.2 | 18 | 2597 | ||
GenBank mRNA | AJ307017.1 | - | 18 | 2597 |