Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zzz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080755.2 | NP_001074224.1 | 4 | 1634 | ||
NM_001287139.1 | NP_001274068.1 | 4 | 1576 | |||
GenBank mRNA | AK034365.1 | - | 1 | 182 | ||
AK131150.1 | - | 4 | 1572 | |||
BC060614.1 | - | 4 | 1566 |