Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144844.2 | NP_659093.2 | 13 | 1215 | ||
GenBank mRNA | AK148002.1 | - | 13 | 1205 | ||
AK188468.1 | - | 2 | 250 | |||
AY046947.1 | - | 13 | 1232 | |||
BC006915.1 | - | 13 | 1212 | 3601148 | ||
BC049802.1 | - | 13 | 1181 | 6332730 |