Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271898.1 | NP_001258827.1 | 10 | 1695 | ||
NM_015729.3 | NP_056544.2 | 10 | 1695 | |||
GenBank mRNA | AB034914.1 | - | 10 | 1384 | ||
AF006688.1 | - | 10 | 1434 | |||
AK034760.1 | - | 6 | 1000 | |||
AK040566.1 | - | 10 | 1398 | |||
AK054446.1 | - | 10 | 1471 | |||
AK128941.1 | - | 10 | 1397 | |||
AK170217.1 | - | 6 | 765 | |||
BC054727.1 | - | 10 | 1363 | |||
BC056448.1 | - | 10 | 1695 | 5704873 |